accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 81633-29-6

IUPAC Name: ethyl 2-amino-4-methylpyrimidine-5-carboxylate
Synonyms: ethyl 2-amino-4-methylpyrimidine-5-carboxylate
Molecular Weight (g/mol): 181.2
Molecular Formula: C8H11N3O2
InChi Key: YBFVMJRSZCVJJP-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN=C(N)N=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results