accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3OPOOCH3H3CCl

CAS RN 81639-99-8

IUPAC Name: bis(2,6-dimethylphenyl) phosphorochloridate
Synonyms: bis(2,6-dimethylphenyl) phosphorochloridate
Molecular Weight (g/mol): 324.74
Molecular Formula: C16H18ClO3P
InChi Key: WXCXJSOCIOHPLT-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C)=C1OP(Cl)(=O)OC1=C(C)C=CC=C1C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results