Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2,6-dimethylphenyl) Chlorophosphate 93.0+%, TCI America™
SDP

Catalog No. B192225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B192225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B192225G Supplier TCI America Supplier No. B192225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 81639-99-8
Molecular Formula C16H18ClO3P
Molecular Weight (g/mol) 324.741
MDL Number MFCD00623313
InChI Key WXCXJSOCIOHPLT-UHFFFAOYSA-N
Synonym Bis(2,6-dimethylphenyl)phosphoryl Chloride, Chlorophosphoric Acid Bis(2,6-dimethylphenyl) Ester
PubChem CID 9923313
IUPAC Name 2-[chloro-(2,6-dimethylphenoxy)phosphoryl]oxy-1,3-dimethylbenzene
SMILES CC1=C(C(=CC=C1)C)OP(=O)(OC2=C(C=CC=C2C)C)Cl

Specifications

Melting Point 52°C
Color White-Yellow
Boiling Point 219°C
Quantity 25 g
UN Number 1759
Formula Weight 324.74
Percent Purity ≥93.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(2,6-dimethylphenyl) Chlorophosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.