accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHONOHOOHO

CAS RN 817-11-8

IUPAC Name: 3-[bis(2-carboxyethyl)amino]propanoic acid
Synonyms: 3-[bis(2-carboxyethyl)amino]propanoic acid
Molecular Weight (g/mol): 233.22
Molecular Formula: C9H15NO6
InChi Key: IWTIBPIVCKUAHK-UHFFFAOYSA-N
SMILES: OC(=O)CCN(CCC(O)=O)CCC(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results