Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,3',3″-Nitrilotripropionic Acid 99.0+%, TCI America™
SDP

Catalog No. N040925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N040925G 25 g
N040910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N040925G Supplier TCI America Supplier No. N040925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 817-11-8
Molecular Formula C9H15NO6
Molecular Weight (g/mol) 233.22
MDL Number MFCD00055827
InChI Key IWTIBPIVCKUAHK-UHFFFAOYSA-N
Synonym 3,3',3-nitrilotripropionic acid, nitrilotripropionic acid, 3,3',3-nitrilotripropanoic acid, beta-alanine, n,n-bis 2-carboxyethyl, 3-bis 2-carboxyethyl amino propanoic acid, acmc-209pm3, 3,3',3-nitrilotripropanoic acid non-preferred name, tris 3-oxo-3-hydroxypropyl amine
PubChem CID 98537
IUPAC Name 3-[bis(2-carboxyethyl)amino]propanoic acid
SMILES C(CN(CCC(=O)O)CCC(=O)O)C(=O)O

Specifications

Color White
Quantity 25 g
Formula Weight 233.22
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,3′,3′′-Nitrilotripropionic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.