accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 817-95-8

IUPAC Name: ethyl 2-ethoxyacetate
Synonyms: ethyl 2-ethoxyacetate
Molecular Weight (g/mol): 132.16
Molecular Formula: C6H12O3
InChi Key: CKSRFHWWBKRUKA-UHFFFAOYSA-N
SMILES: CCOCC(=O)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results