accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH3H3CH3CCH3OOH3C

CAS RN 81749-14-6

IUPAC Name: 1,3-diethyl 2,2-bis(2-methylpropyl)propanedioate
Synonyms: 1,3-diethyl 2,2-bis(2-methylpropyl)propanedioate
Molecular Weight (g/mol): 272.39
Molecular Formula: C15H28O4
InChi Key: NIXFNZVGGMZGPZ-UHFFFAOYSA-N
SMILES: CCOC(=O)C(CC(C)C)(CC(C)C)C(=O)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)