Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl Diisobutylmalonate 98.0+%, TCI America™
SDP

Catalog No. D228025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D228025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D228025ML Supplier TCI America Supplier No. D228025ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 81749-14-6
Molecular Formula C15H28O4
Molecular Weight (g/mol) 272.39
MDL Number MFCD00191422
InChI Key NIXFNZVGGMZGPZ-UHFFFAOYSA-N
Synonym Diisobutylmalonic Acid Diethyl Ester
PubChem CID 10423387
IUPAC Name 1,3-diethyl 2,2-bis(2-methylpropyl)propanedioate
SMILES CCOC(=O)C(CC(C)C)(CC(C)C)C(=O)OCC

Specifications

Color Colorless
Boiling Point 102°C
Quantity 25 mL
Formula Weight 272.39
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Diethyl Diisobutylmalonate
EINECSNumber (2)-0879
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.