accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 818-23-5

IUPAC Name: pentadecan-8-one
Synonyms: 8-pentadecanone
Molecular Weight (g/mol): 226.4
Molecular Formula: C15H30O
InChi Key: PQYGSSYFJIJDFK-UHFFFAOYSA-N
SMILES: CCCCCCCC(=O)CCCCCCC
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)