Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Pentadecanone, 98%

Catalog No. AC169790250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169790250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC169790250 Supplier Thermo Scientific Chemicals Supplier No. 169790250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 818-23-5 | C15H30O | 226.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 818-23-5
Molecular Formula C15H30O
Molecular Weight (g/mol) 226.40
MDL Number MFCD00009555
InChI Key PQYGSSYFJIJDFK-UHFFFAOYSA-N
Synonym 8-pentadecanone, caprylone, heptyl ketone, diheptyl ketone, di-n-heptyl ketone, 8-oxopentadecane, pentadecane-8-one, saturated fatty acid, acmc-1bl8g, caprylone_gurudeebansatyavani
PubChem CID 13162
IUPAC Name pentadecan-8-one
SMILES CCCCCCCC(=O)CCCCCCC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 40°C to 44°C
Color White to Yellow
Boiling Point 178°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3(CH2)6CO(CH2)6CH3
Quantity 25 g
Beilstein 01, 717
Formula Weight 226.4
Percent Purity 98%
Physical Form Shiny Crystals or Flakes
Chemical Name or Material 8-Pentadecanone, 98%
Show More Show Less
EINECSNumber 212-450-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.