accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOHO

CAS RN 818-88-2

IUPAC Name: 10-methoxy-10-oxodecanoic acid
Synonyms: 10-methoxy-10-oxodecanoic acid
Molecular Weight (g/mol): 216.28
Molecular Formula: C11H20O4
InChi Key: OSYQOBUUFRGFNG-UHFFFAOYSA-N
SMILES: COC(=O)CCCCCCCCC(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results