Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monomethyl Sebacate 98.0+%, TCI America™
SDP

Catalog No. S039525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S039525G 25 g
S03955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S039525G Supplier TCI America Supplier No. S039525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 818-88-2
Molecular Formula C11H20O4
Molecular Weight (g/mol) 216.28
MDL Number MFCD00004439
InChI Key OSYQOBUUFRGFNG-UHFFFAOYSA-N
Synonym Decanedioic Acid Monomethyl Ester, Methyl Hydrogen Sebacate, Sebacic Acid Monomethyl Ester
PubChem CID 69957
IUPAC Name 10-methoxy-10-oxodecanoic acid
SMILES COC(=O)CCCCCCCCC(O)=O

Specifications

Melting Point 42°C
Color White
Quantity 25 g
Formula Weight 216.28
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Monomethyl Sebacate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.