accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONCH3NH(E/Z)

CAS RN 81998-07-4

IUPAC Name: 4,4'-dimethyl-1'H,3H-[2,2'-bipyridinyliden]-3-one
Synonyms: 4,4'-dimethyl-1'H-[2,2'-bipyridinyliden]-3-one
Molecular Weight (g/mol): 200.24
Molecular Formula: C12H12N2O
InChi Key: AWUFFAVGWFOSNV-UHFFFAOYSA-N
SMILES: CC1=CC(NC=C1)=C1N=CC=C(C)C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)