Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxy-4,4'-dimethyl-2,2'-bipyridyl 98.0+%, TCI America™
SDP

Catalog No. H1556200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1556200MG 200 mg
H15561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H1556200MG Supplier TCI America Supplier No. H1556200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 81998-07-4
Molecular Formula C12H12N2O
Molecular Weight (g/mol) 200.241
MDL Number MFCD28339195
InChI Key AWUFFAVGWFOSNV-KHPPLWFESA-N
Synonym 4,4′C-Dimethyl-(2,2′C-Bipyridin)-3-ol
PubChem CID 118798227
IUPAC Name (2Z)-4-methyl-2-(4-methyl-1H-pyridin-2-ylidene)pyridin-3-one
SMILES CC1=CC(=C2C(=O)C(=CC=N2)C)NC=C1

Specifications

Melting Point 110°C
Color Yellow
Quantity 200 mg
Formula Weight 200.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxy-4,4′-dimethyl-2,2′-bipyridyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.