accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHOO

CAS RN 82-38-2

IUPAC Name: 1-(methylamino)-9,10-dihydroanthracene-9,10-dione
Synonyms: disperse red 9
Molecular Weight (g/mol): 237.26
Molecular Formula: C15H11NO2
InChi Key: SVTDYSXXLJYUTM-UHFFFAOYSA-N
SMILES: CNC1=CC=CC2=C1C(=O)C1=CC=CC=C1C2=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results