Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Methylamino)anthraquinone 98.0+%, TCI America™
SDP

Catalog No. M0255500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0255500G 500 g
M025525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0255500G Supplier TCI America Supplier No. M0255500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 82-38-2
Molecular Formula C15H11NO2
Molecular Weight (g/mol) 237.258
MDL Number MFCD00001197
InChI Key SVTDYSXXLJYUTM-UHFFFAOYSA-N
PubChem CID 6706
IUPAC Name 1-(methylamino)anthracene-9,10-dione
SMILES CNC1=CC=CC2=C1C(=O)C3=CC=CC=C3C2=O

Specifications

Melting Point 169°C
Quantity 500 g
Formula Weight 237.26
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(Methylamino)anthraquinone
EINECSNumber (4)-0708
RTECSNumber CB0536600
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.