accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 82-46-2

IUPAC Name: 1,5-dichloro-9,10-dihydroanthracene-9,10-dione
Synonyms: 1,5-dichloroanthracene-9,10-dione
Molecular Weight (g/mol): 277.1
Molecular Formula: C14H6Cl2O2
InChi Key: MQIUMARJCOGCIM-UHFFFAOYSA-N
SMILES: ClC1=CC=CC2=C1C(=O)C1=CC=CC(Cl)=C1C2=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)