Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Dichloroanthraquinone, 96%

Catalog No. AAA1953430
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1953430 250 g
AAA1953418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1953430 Supplier Thermo Scientific Chemicals Supplier No. A1953430
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 82-46-2 | C14H6Cl2O2 | 277.1 g/mol

1,5-Dichloroanthraquinone undergoes a solid-state transformation at 470 K. The two modifications are very similar, except that a is greater than b above the transition temperature and the inverse is at room temperature.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Dichloroanthraquinone undergoes a solid-state transformation at 470 K. The two modifications are very similar, except that a is greater than b above the transition temperature and the inverse is at room temperature.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82-46-2
Molecular Formula C14H6Cl2O2
Molecular Weight (g/mol) 277.1
MDL Number MFCD00001190
InChI Key MQIUMARJCOGCIM-UHFFFAOYSA-N
Synonym 1,5-dichloroanthraquinone, 9,10-anthracenedione, 1,5-dichloro, 1,5-dichloro-9,10-anthraquinone, 1,5-dichloranthrachinon, unii-s335v6mf9e, anthraquinone, 1,5-dichloro, 1,5-dichloranthrachinon czech, 1, 5-dichloroanthraquinone, 1,5-dichloroanthra-9,10-quinone, 1,5 dichloro aq
PubChem CID 6711
IUPAC Name 1,5-dichloroanthracene-9,10-dione
SMILES C1=CC2=C(C(=C1)Cl)C(=O)C3=C(C2=O)C(=CC=C3)Cl

Specifications

Melting Point 245°C to 249°C
Quantity 250 g
Beilstein 402592
Solubility Information Insoluble in water.
Formula Weight 277.11
Percent Purity 96%
Chemical Name or Material 1,5-Dichloroanthraquinone
Hazard Category Warning
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.
EINECSNumber 201-424-0
RTECSNumber CB6495000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.