accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 82-57-5

IUPAC Name: 4-methoxy-7-methyl-5H-furo[3,2-g]chromen-5-one
Synonyms: visnagin visnagidin desmethoxykhellin visnagine
Molecular Weight (g/mol): 230.22
Molecular Formula: C13H10O4
InChi Key: NZVQLVGOZRELTG-UHFFFAOYSA-N
SMILES: COC1=C2C=COC2=CC2=C1C(=O)C=C(C)O2
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results