Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methoxy-7-methyl-5H-furo[3,2-g][1]benzopyran-5-one, 97%, Thermo Scientific Chemicals

Catalog No. AC235030250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC235030250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC235030250 Supplier Thermo Scientific Chemicals Supplier No. 235030250

missing translation for 'validMainframeMsg'

CAS: 82-57-5 | Molecular Formula: C13H10O4 | Molecular Weight: 230.219 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82-57-5
MDL Number MFCD00005008
InChI Key NZVQLVGOZRELTG-UHFFFAOYSA-N
Synonym visnagin, desmethoxykhellin, visnagidin, visnagine, 5-methoxy-2-methylfuranochromone, 4-methoxy-7-methyl-5h-furo 3,2-g chromen-5-one, unii-p64438mlbw, 4-methoxy-7-methyl-furo 3,2-g chromen-5-one, chembl45176, 4-methoxy-7-methyl-5h-furo 3,2-g 1-benzopyran-5-one
PubChem CID 6716
ChEBI CHEBI:10002
IUPAC Name 4-methoxy-7-methylfuro[3,2-g]chromen-5-one
SMILES CC1=CC(=O)C2=C(O1)C=C3C(=C2OC)C=CO3

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 139°C to 142°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C13H10O4
Quantity 25 g
Merck Index 15, 10209
Molecular Weight (g/mol) 230.219
Formula Weight 230.22
Percent Purity ≥96.0%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Methoxy-7-methyl-5H-furo[3, 2-g][1]benzopyran-5-one, 97%
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-430-3
RTECSNumber LV1420000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.