accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClH3CNCH3CNCH3H3CNCH3H3CCH3

CAS RN 82-94-0

IUPAC Name: bis[4-(dimethylamino)phenyl][4-(trimethylazaniumyl)phenyl]methylium dichloride
Synonyms: bis[4-(dimethylamino)phenyl][4-(trimethylammonio)phenyl]methylium dichloride
Molecular Weight (g/mol): 458.47
Molecular Formula: C26H33Cl2N3
InChi Key: DWCZIOOZPIDHAB-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].CN(C)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)[N+](C)(C)C
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results