Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Green, TCI America™
SDP

Catalog No. M049825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M049825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M049825G Supplier TCI America Supplier No. M049825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 82-94-0
Molecular Formula C26H33Cl2N3
Molecular Weight (g/mol) 458.471
MDL Number MFCD00011879
InChI Key DWCZIOOZPIDHAB-UHFFFAOYSA-L
Synonym Basic Blue 20
PubChem CID 6727
ChEBI CHEBI:87649
IUPAC Name [4-[[4-(dimethylamino)phenyl]-(4-dimethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]phenyl]-trimethylazanium;dichloride
SMILES CN(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)[N+](C)(C)C.[Cl-].[Cl-]

Specifications

Color Black-Green
Quantity 25 g
Formula Weight 458.47
Physical Form Crystalline Powder
Chemical Name or Material Methyl Green
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.