accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OPBr

CAS RN 82105-88-2

IUPAC Name: [(4-ethoxyphenyl)methyl]triphenylphosphanium bromide
Synonyms: [(4-ethoxyphenyl)methyl]triphenylphosphanium bromide
Molecular Weight (g/mol): 477.38
Molecular Formula: C27H26BrOP
InChi Key: MZBKKJRCQNVENM-UHFFFAOYSA-M
SMILES: [Br-].CCOC1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results