accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 82137-81-3

IUPAC Name: 2'-[(2-chlorophenyl)amino]-6'-(dibutylamino)-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Synonyms: 2'-[(2-chlorophenyl)amino]-6'-(dibutylamino)spiro[2-benzofuran-1,9'-xanthen]-3-one
Molecular Weight (g/mol): 553.1
Molecular Formula: C34H33ClN2O3
InChi Key: ZXYPDMNPLWGWBI-UHFFFAOYNA-N
SMILES: CCCCN(CCCC)C1=CC2=C(C=C1)C1(OC(=O)C3=CC=CC=C13)C1=C(O2)C=CC(NC2=CC=CC=C2Cl)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results