Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-(2-Chloroanilino)-6'-(dibutylamino)fluoran 97.0+%, TCI America™
SDP

Catalog No. C1946500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1946500G 500 g
C194625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1946500G Supplier TCI America Supplier No. C1946500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 82137-81-3
Molecular Formula C34H33ClN2O3
Molecular Weight (g/mol) 553.10
MDL Number MFCD08276326
InChI Key ZXYPDMNPLWGWBI-UHFFFAOYNA-N
PubChem CID 13668872
IUPAC Name 2'-[(2-chlorophenyl)amino]-6'-(dibutylamino)-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
SMILES CCCCN(CCCC)C1=CC2=C(C=C1)C1(OC(=O)C3=CC=CC=C13)C1=C(O2)C=CC(NC2=CC=CC=C2Cl)=C1

Specifications

Melting Point 186°C
Color Red-White
Quantity 500 g
Synonym 2′-(2-Chlorophenylamino)-6′-(dibutylamino)fluoran
Formula Weight 553.10
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2′-(2-Chloroanilino)-6′-(dibutylamino)fluoran
EINECSNumber (4)-1408&(5)-5678
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.