CAS RN 82192-57-2
CAS RN 82192-57-2
IUPAC Name:
(prop-1-yn-1-yl)tris(propan-2-yl)silane
Synonyms:
triisopropyl(prop-1-yn-1-yl)silane
Molecular Weight (g/mol):
196.41
Molecular Formula:
C12H24Si
InChi Key:
FDEZWWXTHRGNJD-UHFFFAOYSA-N
SMILES:
CC#C[Si](C(C)C)(C(C)C)C(C)C
1
–
1
of
1
results
1
1-(Triisopropylsilyl)-1-propyne 95.0+%, TCI America™

CAS: 82192-57-2 Molecular Formula: C12H24Si Molecular Weight (g/mol): 196.409 MDL Number: MFCD00015636 InChI Key: FDEZWWXTHRGNJD-UHFFFAOYSA-N PubChem CID: 4074566 IUPAC Name: tri(propan-2-yl)-prop-1-ynylsilane SMILES: CC#C[Si](C(C)C)(C(C)C)C(C)C