Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Triisopropylsilyl)-1-propyne 95.0+%, TCI America™
SDP

Catalog No. T16905ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16905ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T16905ML Supplier TCI America Supplier No. T16905ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 82192-57-2
Molecular Formula C12H24Si
Molecular Weight (g/mol) 196.409
MDL Number MFCD00015636
InChI Key FDEZWWXTHRGNJD-UHFFFAOYSA-N
PubChem CID 4074566
IUPAC Name tri(propan-2-yl)-prop-1-ynylsilane
SMILES CC#C[Si](C(C)C)(C(C)C)C(C)C

Specifications

Color Yellow
Boiling Point 70°C
Quantity 5 mL
Formula Weight 196.41
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(Triisopropylsilyl)-1-propyne
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.