accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 822-06-0

IUPAC Name: 1,6-diisocyanatohexane
Synonyms: hexamethylene diisocyanate 1,6-hexylene diisocyanate HDI 1,6-hexanediol diisocyanate
Molecular Weight (g/mol): 168.2
Molecular Formula: C8H12N2O2
InChi Key: RRAMGCGOFNQTLD-UHFFFAOYSA-N
SMILES: O=C=NCCCCCCN=C=O
Boiling Point 
  • (5)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (8)
Percent Purity 
  • (2)
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (5)
  • (5)
  • (3)
  • (2)
  • (2)
  • (8)
  • (2)
  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (8)