Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hexamethylene diisocyanate

Catalog No. 52650250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-650-250ML 250 mL
52-650-1L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 52-650-250ML Supplier Sigma Aldrich Supplier No. 52650250ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hexamethylene diisocyanate is an aliphatic diisocyanate monomer typically used to produce oligomers and prepolymers that when combined with a polyol produce light-stable polyurethane.Highly reactive 1,6-hexamethylene diisocyanate (HMDI) was used to synthesize lactic acid polymers from oligomers by the addition of 2,2G-bis(2-oxazoline) (BOX) as chain extenders. Self-healing ability was rendered to polyurethane elastomer by synthesizing alkoxyamine-based diol and reacting it with tri-functional homopolymer of HMDI and polyethylene glycol (PEG). Plastic optical fiber (POF) was prepared by the bulk homopolymerization of HMDI catalyzed by Tin(II)-2 ethylhexanoate (SnOct).

Specifications

CAS 822-06-0
Density 1.047 g/mL (at 20°C)
Boiling Point 82°C to 85°C (0.1 mmHg)
Molecular Formula C8H12N2O2
Refractive Index n20/D 1.453
Linear Formula OCN(CH2)6 NCO
Quantity 250 mL
MDL Number MFCD00002047
Synonym 1,6-Diisocyanatohexane
Molecular Weight (g/mol) 168.19
Percent Purity ≥98.0% (GC)
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.