CAS RN 823-18-7
IUPAC Name:
cyclohexane-1,3-diol
Synonyms:
cyclohexane-1,3-diol
Molecular Weight (g/mol):
116.16
Molecular Formula:
C6H12O2
InChi Key:
RLMGYIOTPQVQJR-UHFFFAOYNA-N
SMILES:
OC1CCCC(O)C1
1
–
1
of
1
results
1
cis-1,3-Cyclohexanediol 98.0+%, TCI America™
CAS: 823-18-7 Molecular Formula: C6H12O2 Molecular Weight (g/mol): 116.16 InChI Key: RLMGYIOTPQVQJR-OLQVQODUSA-N Synonym: cis-1,3-Dihydroxycyclohexane PubChem CID: 6432741 IUPAC Name: (1S,3R)-cyclohexane-1,3-diol SMILES: C1CC(CC(C1)O)O