Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-1,3-Cyclohexanediol 98.0+%, TCI America™
SDP

Catalog No. C23225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23225G 5 g
C23221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C23225G Supplier TCI America Supplier No. C23225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 823-18-7
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
InChI Key RLMGYIOTPQVQJR-OLQVQODUSA-N
Synonym cis-1,3-Dihydroxycyclohexane
PubChem CID 6432741
IUPAC Name (1S,3R)-cyclohexane-1,3-diol
SMILES C1CC(CC(C1)O)O

Specifications

Melting Point 83°C
Color White-Yellow
Boiling Point 125°C
Quantity 5 g
Formula Weight 116.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material cis-1,3-Cyclohexanediol
EINECSNumber (3)-2310
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.