accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 823-69-8

IUPAC Name: 7,7-dichlorobicyclo[4.1.0]heptane
Synonyms: 7,7-dichlorobicyclo[4.1.0]heptane
Molecular Weight (g/mol): 165.06
Molecular Formula: C7H10Cl2
InChi Key: MBPSCGRETWPLSI-UHFFFAOYNA-N
SMILES: ClC1(Cl)C2CCCCC12
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results