Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

7,7-Dichlorobicyclo[4.1.0]heptane 98.0+%, TCI America™
SDP

Catalog No. D43145G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43145G 5 g
D431425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43145G Supplier TCI America Supplier No. D43145G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 823-69-8
Molecular Formula C7H10Cl2
Molecular Weight (g/mol) 165.057
MDL Number MFCD00466186
InChI Key MBPSCGRETWPLSI-UHFFFAOYSA-N
PubChem CID 244375
IUPAC Name 7,7-dichlorobicyclo[4.1.0]heptane
SMILES C1CCC2C(C1)C2(Cl)Cl

Specifications

Color Yellow
Boiling Point 80°C
Quantity 5 g
Formula Weight 165.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 7,7-Dichlorobicyclo[4.1.0]heptane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.