accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CNOCH3

CAS RN 823218-99-1

IUPAC Name: 5-(2,4,6-trimethylphenyl)-3-oxa-4-azatricyclo[5.2.1.0²,⁶]dec-4-ene
Synonyms: 5-(2,4,6-trimethylphenyl)-3-oxa-4-azatricyclo[5.2.1.0²,⁶]dec-4-ene
Molecular Weight (g/mol): 255.36
Molecular Formula: C17H21NO
InChi Key: WKLZNTYMDOPBSE-UHFFFAOYNA-N
SMILES: CC1=CC(C)=C(C2=NOC3C4CCC(C4)C23)C(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results