CAS RN 824-55-5
IUPAC Name:
1-(chloromethyl)-2,4-dimethylbenzene
Synonyms:
1-(chloromethyl)-2,4-dimethylbenzene
Molecular Weight (g/mol):
154.64
Molecular Formula:
C9H11Cl
InChi Key:
BETNPSBTDMBHCZ-UHFFFAOYSA-N
SMILES:
CC1=CC=C(CCl)C(C)=C1
1
–
1
of
1
results
1
2,4-Dimethylbenzyl Chloride (contains 2,6-isomer) 80.0+%, TCI America™
CAS: 824-55-5 Molecular Formula: C9H11Cl Molecular Weight (g/mol): 154.64 MDL Number: MFCD00015556 InChI Key: BETNPSBTDMBHCZ-UHFFFAOYSA-N Synonym: 2,4-dimethylbenzyl chloride,1-chloromethyl-2,4-dimethylbenzene,4-chloromethyl-m-xylene,benzene, 1-chloromethyl-2,4-dimethyl,2,4-dimethyl-1-chloromethyl benzene,1-chloromethyl-2,4-dimethyl-benzene,acmc-209tf3,timtec-bb sbb005790,labotest-bb ltbb004402 PubChem CID: 69989 IUPAC Name: 1-(chloromethyl)-2,4-dimethylbenzene SMILES: CC1=CC=C(CCl)C(C)=C1