Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dimethylbenzyl Chloride (contains 2,6-isomer) 80.0+%, TCI America™
SDP

Catalog No. D028825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D028825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D028825G Supplier TCI America Supplier No. D028825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 824-55-5
Molecular Formula C9H11Cl
Molecular Weight (g/mol) 154.64
MDL Number MFCD00015556
InChI Key BETNPSBTDMBHCZ-UHFFFAOYSA-N
Synonym 2,4-dimethylbenzyl chloride, 1-chloromethyl-2,4-dimethylbenzene, 4-chloromethyl-m-xylene, benzene, 1-chloromethyl-2,4-dimethyl, 2,4-dimethyl-1-chloromethyl benzene, 1-chloromethyl-2,4-dimethyl-benzene, acmc-209tf3, timtec-bb sbb005790, labotest-bb ltbb004402
PubChem CID 69989
IUPAC Name 1-(chloromethyl)-2,4-dimethylbenzene
SMILES CC1=CC=C(CCl)C(C)=C1

Specifications

Color Colorless
Boiling Point 105°C
Quantity 25 g
UN Number 3265
Formula Weight 154.64
Percent Purity ≥80.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Dimethylbenzyl Chloride (contains 2,6-isomer)
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.