CAS RN 824395-67-7
IUPAC Name:
1-[2-(2,5-diphenylphospholan-1-yl)ethyl]-2,5-diphenylphospholane
Synonyms:
1-[2-(2,5-diphenylphospholan-1-yl)ethyl]-2,5-diphenylphospholane
Molecular Weight (g/mol):
506.61
Molecular Formula:
C34H36P2
InChi Key:
VHHAZLMVLLIMHT-UHFFFAOYNA-N
SMILES:
C(CP1C(CCC1C1=CC=CC=C1)C1=CC=CC=C1)P1C(CCC1C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Sigma Aldrich (+)-1,2-Bis((2S,5S)-2,5-Diphenylphospholano)Ethane
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.