accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NBrHNNN

CAS RN 82499-03-4

IUPAC Name: 6-bromo-7H-purin-2-amine
Synonyms: 6-bromo-7H-purin-2-amine
Molecular Weight (g/mol): 214.03
Molecular Formula: C5H4BrN5
InChi Key: HPGBGNVPUMCKPM-UHFFFAOYSA-N
SMILES: NC1=NC(Br)=C2NC=NC2=N1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)