accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOH3CCH3H3CHO

CAS RN 82578-45-8

IUPAC Name: tert-butyl 3-hydroxybutanoate
Synonyms: tert-butyl 3-hydroxybutanoate
Molecular Weight (g/mol): 160.21
Molecular Formula: C8H16O3
InChi Key: PKPVFILAHLKSNJ-UHFFFAOYNA-N
SMILES: CC(O)CC(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results