accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClCH3H3COH3CH3CHN

CAS RN 826-36-8

IUPAC Name: hydrogen 2,2,6,6-tetramethylpiperidin-4-one chloride
Synonyms: triacetone amine
Molecular Weight (g/mol): 191.7
Molecular Formula: C9H18ClNO
InChi Key: ZXNWYMNKYXUZGM-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1(C)CC(=O)CC(C)(C)N1
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)