accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 826-39-1

IUPAC Name: hydrogen N,2,3,3-tetramethylbicyclo[2.2.1]heptan-2-amine chloride
Synonyms: hydrogen mecamylamine chloride
Molecular Weight (g/mol): 203.75
Molecular Formula: C11H22ClN
InChi Key: PKVZBNCYEICAQP-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CNC1(C)C2CCC(C2)C1(C)C
Color 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results