accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 826-73-3

O

CAS RN 826-73-3

IUPAC Name: 6,7,8,9-tetrahydro-5H-benzo[7]annulen-5-one
Synonyms: 6,7,8,9-tetrahydrobenzo[7]annulen-5-one
Molecular Weight (g/mol): 160.22
Molecular Formula: C11H12O
InChi Key: KWHUHTFXMNQHAA-UHFFFAOYSA-N
SMILES: O=C1CCCCC2=CC=CC=C12
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results