accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 82671-02-1

IUPAC Name: 2,6-dichloro-5-fluoropyridine-3-carbonitrile
Synonyms: 2,6-dichloro-5-fluoropyridine-3-carbonitrile
Molecular Weight (g/mol): 190.99
Molecular Formula: C6HCl2FN2
InChi Key: DEDKKOOGYIMMBC-UHFFFAOYSA-N
SMILES: FC1=CC(C#N)=C(Cl)N=C1Cl
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)