Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloro-5-fluoro-3-pyridinecarbonitrile 98.0+%, TCI America™
SDP

Catalog No. D39725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D39725G Supplier TCI America Supplier No. D39725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 82671-02-1 | C6HCl2FN2 | 190.986 g/mol

Chemical Identifiers

CAS 82671-02-1
Molecular Formula C6HCl2FN2
Molecular Weight (g/mol) 190.986
MDL Number MFCD01863289
InChI Key DEDKKOOGYIMMBC-UHFFFAOYSA-N
Synonym 2,6-dichloro-5-fluoro-3-pyridinecarbonitrile, 2,6-dichloro-5-fluoronicotinonitrile, 2,6-dichloro-3-cyano-5-fluoropyridine, 3-cyano-2,6-dichloro-5-fluoropyridine, 2,6-dichloro-5-fluoro-nicotinonitrile, 2,6-dichloro-5-fluoro-3-cyanopyridine, 2,6-dichloro-5-fluoro-pyridine-3-carbonitrile, 3-pyridinecarbonitrile, 2,6-dichloro-5-fluoro, pubchem10945, acmc-209ppr
PubChem CID 2733849
IUPAC Name 2,6-dichloro-5-fluoropyridine-3-carbonitrile
SMILES C1=C(C(=NC(=C1F)Cl)Cl)C#N

Specifications

Melting Point 91°C
Quantity 5 g
UN Number 3439
Formula Weight 190.99
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dichloro-5-fluoro-3-pyridinecarbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.