CAS RN 827343-08-8
IUPAC Name:
2-bromo-3-hexadecylthiophene
Synonyms:
2-bromo-3-hexadecylthiophene
Molecular Weight (g/mol):
387.46
Molecular Formula:
C20H35BrS
InChi Key:
GOHORSZVGYZIGO-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCCC1=C(Br)SC=C1
1
–
1
of
1
results
1
2-Bromo-3-hexadecylthiophene 96.0+%, TCI America™
CAS: 827343-08-8 Molecular Formula: C20H35BrS Molecular Weight (g/mol): 387.46 MDL Number: MFCD30742896 InChI Key: GOHORSZVGYZIGO-UHFFFAOYSA-N PubChem CID: 69331498 IUPAC Name: 2-bromo-3-hexadecylthiophene SMILES: CCCCCCCCCCCCCCCCC1=C(Br)SC=C1