Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-3-hexadecylthiophene 96.0+%, TCI America™
SDP

Catalog No. B54091G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B54091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B54091G Supplier TCI America Supplier No. B54091G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 827343-08-8
Molecular Formula C20H35BrS
Molecular Weight (g/mol) 387.46
MDL Number MFCD30742896
InChI Key GOHORSZVGYZIGO-UHFFFAOYSA-N
PubChem CID 69331498
IUPAC Name 2-bromo-3-hexadecylthiophene
SMILES CCCCCCCCCCCCCCCCC1=C(Br)SC=C1

Specifications

Melting Point 27°C
Color White
Quantity 1 g
Formula Weight 387.46
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-3-hexadecylthiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.