CAS RN 827614-64-2
IUPAC Name:
5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
Synonyms:
5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
Molecular Weight (g/mol):
220.08
Molecular Formula:
C11H17BN2O2
InChi Key:
YFTAUNOLAHRUIE-UHFFFAOYSA-N
SMILES:
CC1(C)OB(OC1(C)C)C1=CC=C(N)N=C1
1
–
1
of
1
results
1
Sigma Aldrich 2-Aminopyridine-5-Boronic Acid Pinacol Ester
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.