accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 828-26-2

IUPAC Name: hexamethyl-1,3,5-trithiane
Synonyms: hexamethyl-1,3,5-trithiane
Molecular Weight (g/mol): 222.42
Molecular Formula: C9H18S3
InChi Key: NBNWHQAWKFYFKI-UHFFFAOYSA-N
SMILES: CC1(C)SC(C)(C)SC(C)(C)S1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)