Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,4,4,6,6-Hexamethyl-S-trithiane, 97%, Thermo Scientific™

Catalog No. AC332660050
Change view
Click to view available options
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC332660050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC332660050 Supplier Thermo Scientific Chemicals Supplier No. 332660050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 828-26-2
CAS Min % 96.0
CAS Max % 100.0
Density 1.06g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C9H18S3
Refractive Index 1.5400 to 1.5440 (20°C, 589nm)
Quantity 5g
MDL Number MFCD00066341
Synonym trithioacetone, 2,2,4,4,6,6-hexamethyl-s-trithiane, hexamethyl-1,3,5-trithiane, hexamethyl-s-trithiane, 1,3,5-trithiane, 2,2,4,4,6,6-hexamethyl, s-trithiane, hexamethyl, unii-l0me5b2zwy, fema no. 3475, s-trithiane, 2,2,4,4,6,6-hexamethyl, l0me5b2zwy
InChI Key NBNWHQAWKFYFKI-UHFFFAOYSA-N
SMILES CC1(C)SC(C)(C)SC(C)(C)S1
IUPAC Name hexamethyl-1,3,5-trithiane
Molecular Weight (g/mol) 222.42
PubChem CID 13233
Formula Weight 222.4
Percent Purity ≥96.0%
Chemical Name or Material 2,2,4,4,6,6-Hexamethyl-S-trithiane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-582-5
RTECSNumber YL8350000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.