accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 82846-18-2

IUPAC Name: 4-fluoro-3-methoxybenzoic acid
Synonyms: 4-fluoro-3-methoxybenzoic acid
Molecular Weight (g/mol): 170.14
Molecular Formula: C8H7FO3
InChi Key: LWGCZCMLPRMKIZ-UHFFFAOYSA-N
SMILES: COC1=C(F)C=CC(=C1)C(O)=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)